Would you like to log in again?
PTS Panels® Lipid + eGLU Smart Bundle
Supplier: PTS Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PTS Panels®
2729
76994-110EA 315.1 USD InStock
76994-110
PTS Panels® Lipid + eGLU Smart Bundle
Clinical Diagnostic Supplies Clinical Reagents and Kits
PTS Panels Lipid + eGLU Smart Bundle allows you to get accurate lipid and blood glucose results in as little as 90 seconds, from one fingerstick.
  • For use with the CardioChek Plus analyzer
  • 15-count lipid test strips vial and one 15-count electrochemical glucose (eGLU) test strips vial
  • Accurate results in as little as 90 seconds
PTS Panels Lipid Panel+eGLU test strips are for professional use exclusively with the CardioChek Plus analyzer to measure total cholesterol, high density lipoprotein (HDL) cholesterol, Triglycerides and glucose in venous whole blood or capillary fingerstick blood.
Cholesterol measurements are used in the diagnosis and treatments of disorders involving excess cholesterol in the blood and lipid and lipoprotein metabolism disorders. Lipoprotein measurements are used in the diagnosis and treatment of lipid disorders (such as diabetes mellitus), atherosclerosis, and various liver and renal diseases. Glucose measurements are used in the diagnosis and treatment of carbohydrate metabolism disorders including diabetes mellitus, neonatal hypoglycemia, and idiopathic hypoglycemia, and of pancreatic islet carcinoma.
This test system will display numeric results in the following ranges:
Cholesterol: 100 to 400 mg/dl (2.59 to 10.36 mmol/l)
Hdl Cholesterol: 20 to 120 mg/dl (0.52 to 3.11 mmol/l)
Triglycerides: 50 to 500 mg/dl (0.57 to 5.65 mmol/l)
Glucose: 40 to 600 mg/dl (2.22 to 33.3 mmol/l)
Certifications: CLIA-waived, FDA-cleared.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty