Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Limulus Color KY Series
Supplier: Fujifilm Wako Chemicals USA
291-53101 291-53601
76994-356EA 674.76 USD InStock
76994-356 76994-376
Limulus Color KY Series
Microbiology Test Systems Endotoxin Tests
The Limulus Color KY Series are quantitative kinetic chromogenic assays (KCA) that are endotoxin-specific and unreactive to (1,3)-ß-D-glucan. This series includes both a multi-test kit and a single-test kit, which utilize a synthetic substrate that produces a yellow color to detect endotoxin with high sensitivity.
  • Reagent reacts with endotoxin over a wide range of concentrations
  • Reduces the chance of interfering factors due to high sensitivity
  • Can be used with the Toxinometer® or microplate reader
  • KCA quantitative range detection limit of 0.0002 to 5 EU/ml for single test and 0.0005 to 5 EU/ml for multi test
  • The sample size is 0.1 ml when used with tube reader; 0.05 ml sample size when used with microplate reader
All reagent kits are matched with a Control Standard Endotoxin (CSE). These reagents work well with samples that are very turbid or show difficulty in quantification by the kinetic turbidimetric assay (KTA).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty