Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Zyppy™ Plasmid Purification Kits, Zymo Research
Supplier: Zymo Research
Zyppy™
D4037 D4041 D4042 D4100 D4019 D4101 D4036 D4102 D4043 D4020
77001-390KT 389.8 USD InStock
77001-390 77001-290 77001-302 77001-300 77001-234 77001-288 77001-166 77001-164 77001-394 77001-392
Zyppy™ Plasmid Purification Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Zyppy™ Plasmid Purification Kits feature a pellet-free procedure for the fastest purification of plasmid DNA.
  • The fastest, easiest miniprep available for purifying high-quality plasmid DNA
  • Pellet-free procedure omits conventional cell pelleting and re-suspension steps
  • DNA quality is sufficient for cloning and sequencing
The Zyppy™ Plasmid Kits feature a pellet-free modified alkaline lysis method that bypasses bacterial culture centrifugation and re-suspension steps common to classical plasmid preparation procedures. Simply add the uniquely formulated 7X Lysis Buffer directly to bacterial culture, neutralize, and then purify using the provided Zymo-Spin column technology. Additionally, the innovative coloured buffers included in the kit permit error-free visualization and identification of complete bacterial cell lysis and neutralization. The Zyppy™ Plasmid Kits are the fastest and easiest method available to separate plasmid DNA from E. Coli efficiently. The plasmid DNA is of the highest quality, and is well suited for use in bacterial transformation, restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty