Would you like to log in again?
Genomic DNA Clean & Concentrator™ Kits, Zymo Research
Supplier: Zymo Research
Clean & Concentrator™
D4065 D4064 D4067 D4066 D4010 D4011
77001-346KT 763 USD InStock
77001-346 77001-148 77001-344 77001-342 77001-340 77001-150
Genomic DNA Clean & Concentrator™ Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Quick (5 minute) clean-up of large-sized DNA from any enzymatic reaction or impure preparation without messy precipitations.
  • Quick (5 minute) protocol
  • Unique spin column for low volume (≥ 10 µl) elution of ultra-pure, high-yield DNA
  • Eluted DNA is ideal for PCR, endonuclease digestion, sequencing, and more
The Genomic DNA Clean & Concentrator™ (DCC™) is designed for the quick (5 minute) recovery of ultra-pure, large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, (wga) DNA, etc.) from any enzymatic reaction or impure preparation (e.g., Proteinase K digestion). There is no need for organic denaturants, chloroform, or messy precipitations. Simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied Zymo-Spin™ Column. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. The product is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions, crude plasmid preparations, cDNA synthesis, and more.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty