Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
EZ DNA Methylation™-Direct Kits, Zymo Research
Supplier: Zymo Research
EZ DNA Methylation™
D5044 D5022 D5020 D5045 D5021 D5023
77001-636KT 676.1 USD InStock
77001-636 77001-646 77001-644 77001-682 77001-680 77001-628
EZ DNA Methylation™-Direct Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Complete bisulfite conversion of DNA directly from blood, soft tissue, cells, FFPE samples, and LCM samples.
  • Compatible with small sample inputs - as few as 10 cells or 50 pg DNA
  • Desulphonation and recovery of bisulfite-treated DNA with a spin-column, 96-well plate, or magnetic beads
  • Low fragmentation
  • Includes Proteinase K for tissue digestion
The EZ DNA Methylation-Direct™ Kit is a further refinement of our popular EZ DNA Methylation™ and EZ DNA Methylation-Gold® kits. The EZ DNA Methylation-Direct™ Kit features reliable and complete bisulfite conversion of DNA directly from blood, tissue, and cells without the prerequisite for DNA purification. The increased sensitivity of these kits make it possible to amplify bisulfite-converted DNA from as few as 10 cells or 50 pg DNA. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including restriction endonuclease digestion, sequencing, microarrays, etc.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty