Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DNA STANDARD SET 5-MC 5-HMC 50 NG/UL
Supplier: Zymo Research
5-Methylcytosine / 5-Hydroxymethylcytosine DNA Standard Set^ 5-Methylcytosine / 5-Hydroxymethylcytosine DNA Standard Set, Concentration 50 ng/ul, Includes 3 tubes each containing 2 ug of 100% unmodified cytosine, 5-methylcytosine, and 5-hydroxymethylcytosine DNA standard
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DNA STANDARD SET 5-MC 5-HMC 50 NG/UL 77001-768 D5405 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty