Would you like to log in again?
Quick-DNA™ Fungal/Bacterial Kits, Zymo Research
Supplier: Zymo Research
Quick-DNA™
D6007 D6006 D6005 D6105
77001-808KT 280 USD InStock
77001-808 77001-810 77001-812 77001-836
Quick-DNA™ Fungal/Bacterial Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Quick, efficient isolation of DNA from all types of tough-to-lyse fungi and bacteria.
  • Simple, efficient isolation of DNA from all types of tough-to-lyse fungi and bacteria in minutes
  • Ultra-high density BashingBeads™ are fracture resistant and chemically inert
The Quick-DNA™ Fungal/Bacterial Kits are designed for the simple and rapid isolation of DNA from tough-to-lyse fungi, including A. fumigatus, C. albicans, N. crassa, S. cerevisiae, S. pombe, as well as Gram-positive/negative bacteria, algae, and protozoa. The procedures are easy and can be completed in minutes. Fungal and/or bacterial samples are rapidly and efficiently lysed with our state-of-the-art, ultra-high density BashingBeads™. Zymo-Spin™ technology is then used to isolate the DNA, which is ideal for downstream molecular-based applications including PCR, array, etc.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty