Would you like to log in again?
STANDARD MICROBIAL COMMUNITY ZYMOBIOMICS
Supplier: Zymo Research
ZymoBIOMICS* Microbial Community DNA Standard 200ng^ ZymoBIOMICS* Microbial Community DNA Standard, Source A mixture of genomic DNA of ten microbial strains. Ideal for the validation and optimization, along with the quality control of microbiomics or metagenomcis workflows. Size: 200ng
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STANDARD MICROBIAL COMMUNITY ZYMOBIOMICS 77001-844 D6305 Each (200Ng) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty