Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
His-Spin Protein Kits, Zymo Research
Supplier: Zymo Research
P2003-2 P2001 P2002
77001-992KT 83.8 USD InStock
77001-992 77001-994 77001-996
His-Spin Protein Kits, Zymo Research
Protein Purification Protein Purification Kits
A method for fast His-tagged protein purification.
  • Fast (5 minute) method for the purification of His-tagged proteins from cell free extracts
  • Screen bacterial colonies directly on the basis of protein expression vs. plasmid DNA
  • No special instrumentation is required other than a benchtop microcentrifuge
The His-Spin Protein Miniprep™ provides researchers with a method for fast His-tagged protein purification. The easy-to-follow procedure is based on a nickel-charged His-Affinity Gel (IMAC), innovative protein purification, and unique Zymo-Spin™ Column technology. Up to 1 mg of His-tagged protein can be purified in as little as 5 minutes and can be eluted into as little as 100 μL of the provided His-Elution Buffer. The purified protein can be used directly for enzymatic assays, protein biochemical analyses, SDS-PAGE, as well as other protein based applications. The His-Spin Protein Miniprep™ has been optimized to yield maximal protein purity indices: a single protein band is often visualized following Coomassie Blue staining of proteins in SDS-PAGE gel. The straightforward spin-wash-elute protocol dramatically simplifies protein purification and results are obtained in minutes, not hours!
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty