Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Anti-Cytomegalovirus gB Protein Antibody IgG Titer Serologic Assay Kit

Supplier: ReVacc Scientific
X66D33
77007-450EA 470.25 USD InStock
77007-450
Mouse Anti-Cytomegalovirus gB Protein Antibody IgG Titer Serologic Assay Kit
Assays Immunological Assays
This kit is developed for serologic test for mouse IgG titer of Anti-HCMV gB antibody in serum/plasma or purified mouse antibody (monoclonal and polyclonal) in vitro.

Assay principles: This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti-HCMV (Towne strain) gB mouse IgG in serum. The kit consists of high-bind detachable 96-well plate pre-coated with NP protein, positive control, negative control, an HRP-Anti-Mouse IgG secondary antibody, TMB and dilution/wash/stop buffer.

Use protocol (4 simple steps):
1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 hour or room temperature for 1.5 hour. Wash wells 4 times by wash solution (0.3 ml).
2. Dilute the secondary antibody HRP-Anti-Mouse IgG by wash solution and add to each well (0.1 ml). Incubate at room temperature for 0.5 to 1 hour. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml) and incubate at room temperature for 5 to 30 minutes.
4. Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD value reflects the amount of antibody bound.

Storage and shipping condition: Shipment with room temperature. The unopened kit is stable for 6 months from the date of manufacture when stored at 2 to 8 °C. Avoid light. The shelf life is 60 days from the date of opening.

Caution: It is for research use only.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty