Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AAV Rabbit Monoclonal Antibody [clone: 5G4]

Supplier: Genscript
GenScript®
A02206-40
77007-506EA 355.3 USD InStock
77007-506
Anti-AAV Rabbit Monoclonal Antibody [clone: 5G4]
Antibodies
Adeno-associated virus (AAV) is a single-stranded DNA virus that commonly infects humans without pathology. Therefore, AAV can be engineered to deliver DNA to target cells as a vector for gene therapy. There are several serotypes including AAV type 1, 2, 5, 6, 8, 9, DJ and rh.1. Among them, AAV2 is the best characterized and most commonly used. Cap gene of AAV gives rise to three capsid proteins, VP1, VP2 and VP3, with molecular weight of 87, 72 and 62 kDa, respectively. These capsid proteins assemble into a near-spherical protein shell of 60 subunits for AAV.

  • High specificity
  • High affinity
  • High stability

GenScript’s AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.

Type:
Antigen: AAV
Clonality: Monoclonal
Clone: 5G4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty