Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SARS-CoV-2 Nucleocapsid Human Monoclonal Antibody [clone: N22IgA]

Supplier: Genscript
GenScript®
A02090-1 A02090-100
77007-518EA 429 USD InStock
77007-518 77007-636
Anti-SARS-CoV-2 Nucleocapsid Human Monoclonal Antibody [clone: N22IgA]
Antibodies
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). Nucleocapsid Protein is a most abundant structure protein of the coronavirus which is associated with nucleic acid. SARS-CoV-2 Nucleocapsid Antibody (N22IgA), Human Chimeric is produced from cell culture in vitro under conditions free from animal-derived components.

  • High specificity
  • High affinity
  • High stability

Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards, and control antibodies.

Type:
Antigen: SARS-CoV-2 Nucleocapsid
Clonality: Monoclonal
Clone: N22IgA
Conjugation:
Epitope:
Host: Human
Isotype: IgA
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty