Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AAV2 Rabbit Monoclonal Antibody [clone: 15G4]

Supplier: Genscript
GenScript®
A02202-40
77007-532EA 355.3 USD InStock
77007-532
Anti-AAV2 Rabbit Monoclonal Antibody [clone: 15G4]
Antibodies
Adeno-associated virus type 2 (AAV2) is a valuable vector for gene therapy. It is one member of the human parvovirus family which depends on co-infection with helper viruses such as adenovirus or herpes virus for efficient reproduction. AAV2 empty capsids are composed of three proteins, VP1, VP2 and VP3 with relative molecular masses of 87, 72 and 62 kDa, respectively.

  • High specificity
  • High affinity
  • High stability

GenScript’s AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.

Type:
Antigen: AAV2
Clonality: Monoclonal
Clone: 15G4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty