Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Protein A Mouse Monoclonal Antibody [clone: 4F9A8]

Supplier: Genscript
GenScript®
A01778-40
77007-544EA 106.53 USD InStock
77007-544
Anti-Protein A Mouse Monoclonal Antibody [clone: 4F9A8]
Antibodies
Protein A, a bacterial cell wall protein isolated from Staphylococcus aureus, binds to mammalian IgGs mainly through Fc regions. The Protein A is most commonly bound to a chromatography resin, used for production of antibody drugs or reagents. Protein A antibody can be used for quantification of Protein A in drugs and foods or for evaluation of Protein A resin. This Protein A mAb is advantageous due to its IgG1 isotype, for mouse IgG1 has weak binding capacity to Protein A. GenScript Protein A Antibody, mAb, Mouse is produced from the hybridoma resulting from fusion of SP2/0-Ag14 myeloma and B-lymphocytes obtained from mouse immunized with purified recombinant Protein A.

  • High specificity
  • High affinity
  • High stability

GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.

Type: Primary
Antigen: Protein A
Clonality: Monoclonal
Clone: 4F9A8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty