Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Avelumab Mouse Monoclonal Antibody [clone: 12H7]

Supplier: Genscript
GenScript®
A02121-40
77007-586EA 150.88 USD InStock
77007-586
Anti-Avelumab Mouse Monoclonal Antibody [clone: 12H7]
Antibodies
Avelumab with trade name Bavencio, is an FDA-approved drug to treat Merkel cell carcinoma, urothelial carcinoma, and renal cell carcinoma. Avelumab is a whole monoclonal antibody of isotype IgG1 that binds to the programmed death-ligand 1 (PD-L1) and therefore inhibits binding to its receptor programmed cell death 1 (PD-1). PD-L1 can be highly expressed on certain tumors, which is thought to lead to reduced activation of immune cells (cytotoxic T-cells in particular) that might otherwise recognize and attack the cancer. Inhibition of PD-L1 by Avelumab can remove this inhibitor effect and thereby engender an anti-tumor response. It is one of several ways to block inhibitory signals related to T-cell activation, a more general strategy known as immune checkpoint inhibition. Anti-Avelumab Antibody (12H7), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Avelumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Avelumab
Clonality: Monoclonal
Clone: 12H7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty