Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SARS-CoV-2 Spike protein Human Monoclonal Antibody [clone: S9HC]

Supplier: Genscript
GenScript®
A02133-1 A02133-100
77007-590EA 391.88 USD InStock
77007-590 77007-554
Anti-SARS-CoV-2 Spike protein Human Monoclonal Antibody [clone: S9HC]
Antibodies
SARS-CoV-2 ADE-causing antibody is a SARS-CoV-2 Spike protein antibody that can exacerbate the COVID-19 disease severity of patients. Antibody-dependent enhancement (ADE) may be involved in the clinical observation of patients with increased severity of symptoms and high levels of SARS-CoV-2 antibodies. It is reported [1] that the same antibodies against the NTD domain of the SARS-CoV-2 spike protein have an ADE effect by increasing the binding of ACE2 to the spike protein.

  • High specificity
  • High affinity
  • High stability

Genscript offers site-specific neutralizing antibodies, neutralizing antibody standards, and control antibodies.

Type:
Antigen: SARS-CoV-2 Spike protein
Clonality: Monoclonal
Clone: S9HC
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty