Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti- Mouse Monoclonal Antibody (Biotin) [clone: 2C4]

Supplier: Genscript
GenScript®
A02160-40
77007-596EA 250.8 USD InStock
77007-596
Anti- Mouse Monoclonal Antibody (Biotin) [clone: 2C4]
Antibodies
Adeno-associated virus type 2 (AAV2) is a valuable vector for gene therapy. It is one member of the human parvovirus family which depends on co-infection with helper viruses such as adenovirus or herpes virus for efficient reproduction. The AAV2 capsid (intact particle) comprises three types of subunits, VP1, VP2, and VP3, a total of 60 copies in a ratio of 1:1:8 (VP1:VP2:VP3). For the application of AAV2 vector in gene therapy, the determination of total capsid titer is essential, accurate characterization and quantification of purified AAV2 particle product represent a critical step for clinical application. GenScript Anti-AAV2 (intact particle) Antibody (2C4), mAb, Mouse is produced from cell culture in vitro under conditions free from animal-derived components.

  • High specificity
  • High affinity
  • High stability

GenScript's AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.

Type:
Antigen:
Clonality: Monoclonal
Clone: 2C4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Multiple

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty