Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Dupilumab Mouse Monoclonal Antibody [clone: 2D5]

Supplier: Genscript
GenScript®
A02097-40
77007-604EA 150.88 USD InStock
77007-604
Anti-Dupilumab Mouse Monoclonal Antibody [clone: 2D5]
Antibodies
Dupilumab with trade name Dupixent, is an FDA-approved drug for the treatment of patients aged 6 years and older with moderate-to-severe atopic dermatitis. It is a human monoclonal antibody. Dupilumab binds to the interleukin-4 receptor alpha (IL-4R alpha) subunit shared by the interleukin (IL)-4 and IL-13 receptor complexes, thereby inhibiting IL-4 and IL-13 signaling. Anti-Dupilumab Antibody (2D5), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Dupilumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Dupilumab
Clonality: Monoclonal
Clone: 2D5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty