Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DXd Mouse Monoclonal Antibody [clone: 39F4]

Supplier: Genscript
GenScript®
A02217-40
77007-650EA 167.2 USD InStock
77007-650
Anti-DXd Mouse Monoclonal Antibody [clone: 39F4]
Antibodies
Payloads are the important components of antibody-drug conjugates (ADCs). The most commonly used ADC payloads are MMAE, DM1, and DXd. ADC pharmacokinetic (PK) studies include free payload analysis, conjugated antibody analysis, and total antibody analysis via ligand-binding assay (LBA) and LC-MS/MS assay. The anti-payload antibody is a useful reagent in PK assay for determining conjugated antibodies. It can be used as a capture antibody to determine the conjugated antibody in LBA assay or through immunocapture to enrich the ADC sample in LC-MS/MS assay. The DXd, also known as exatecan derivative, is a potent inhibitor of DNA topoisomerase I. Trastuzumab deruxtecan (T-DXd, DS-8201a or Enhertu®), composed of trastuzumab conjugated to DXd, is the first approved ADC for the treatment of breast cancer.

  • High specificity
  • High affinity
  • High stability

GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.

Type: Primary
Antigen: DXd
Clonality: Monoclonal
Clone: 39F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty