Would you like to log in again?
HydroFoamer HydroSprayer Device
Supplier: Unimed
HydroFoamer HydroSprayer
HPGF412483
77007-796EA 183.28 USD InStock
77007-796
HydroFoamer HydroSprayer Device
Spray Guns
Increase labor productivity with the HydroFoamer and HydroSprayer. These easy-to-use spraying and foaming devices can be utilized anywhere a hose can reach.
  • Proven, reliable components
  • Non-electric
  • No installation required
  • Compact, easy to store
  • Enhanced safety due to reduced exposure to chemicals
  • Maximum cleaning product performance
Chemical concentrates will perform at their peak because they are mixed only as they are applied. Just fill the concentrate container and connect the dispenser to a water source. Quick application, properly diluted chemicals, and minimal waste make the HydroFoamer and HydroSprayer devices some of the most popular products in the industry.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty