Would you like to log in again?
Hydro AccuMax Dilution System, 1-Button Dilution, High Flow
Supplier: Unimed
Hydro AccuMax
HDSC412331
77007-812EA 313.42 USD InStock
77007-812
Hydro AccuMax Dilution System, 1-Button Dilution, High Flow
Spray Guns
Systems using AccuPro pressure regulation technology eliminate dilution variance by controlling the water pressure into the unit, never allowing the dilution to 'lean out' the desired dilution target.
  • 1-button dilution: high flow carboy fill (3.5 gpm)
  • New, expansive workspace to easily route tubes
  • Easy, tool-free maintenance
  • E-gap and AirGap options to satisfy regulatory standards
  • Same mounting brackets as 'Hydro Systems' AccuPro and AccuDose
High water pressure results in improperly diluted chemicals, poor cleaning performance, and lost chemical sales. AccuPro technology delivers accurate dilution regardless of fluctuations in water pressure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty