Would you like to log in again?
Nilfisk Nommo Foam/Rinse/Sanitize Unit
Supplier: Unimed
Nilfisk Nommo
FFRS516MMO
77007-824EA 827.28 USD InStock
77007-824
Nilfisk Nommo Foam/Rinse/Sanitize Unit
Spray Guns
Clean and sanitize small to medium spaces with the water-driven Nommo. Three functions in one unit save space and simplify the sanitation process.
  • Powered by water
  • Draws from two concentrated products, one at a time
  • Color-coded selector valve matches chemical suction lines
  • Control dilution rate for each product independently using metering tips
  • Spray or foam release; switch function by swapping nozzles
  • Simple installation and operation
The unit is powered by water, requiring 29 to 87 psi (2 to 6 bar) of incoming water pressure, and draws from two concentrated products, one at a time.
Choose from two nozzle types; the patented Tornado rinse/spray nozzle provides high-impact output with 20% less water, while the top jet foam nozzle creates visible, wet foam without compressed air.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty