Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lighthouse Foam Unit, 0.4 Gallon

Supplier: Unimed
Lighthouse
FPFU51615L
77007-828EA 100.04 USD InStock
77007-828
Lighthouse Foam Unit, 0.4 Gallon
Spray Guns
This small foaming unit is perfect for small spaces and quick disinfections. Best used in restrooms, vehicles, and other spaces that have targeted disinfection areas.

  • Comfortable grip handle with trigger for quick on/off control
  • Nozzle with 110° fan tip
  • Pressure relief valve for safety
  • Viton pump seals for chemical compatibility
  • Polypropylene tank and pump head
  • View stripe for easy visibility of product levels
  • Stability base with gripping notches

Compact unit with manual pump provides maximum portability for small jobs and spot cleaning.

Color-coding options include black or white finishes.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty