Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
TaskBrand® V40 Value Series DRC Wipes
Supplier: High-Tech Conversions
TaskBrand®
N-V040JPW N-V040IDW2 N-V040QPW N-V040IDB
77007-968CS 82.8 USD InStock
77007-968 77007-914 77008-060 77007-834
TaskBrand® V40 Value Series DRC Wipes
Wipes Disposable Wipes and Towels Wipes for General Applications
DRC is the leading all-purpose disposable wipe.
  • All-purpose disposable wipe
  • Highly absorbent dry wipe
  • Heavy-duty, low linting
  • Unique stretch with cloth-like feel
  • Made of 35% (avg.) post-industrial recycled fibers
TaskBrand® V40 value series DRC wipes are made of 35% post-industrial recycled fibers, making up a minimum 85% of the total product weight. Applications include glass and mirror cleaning, equipment/tools/parts cleaning, staining, wiping up oil/grease/water, general-purpose surface and hand cleaning, janitorial and facility maintenance applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty