Would you like to log in again?
ULTIMATE 60™ Sealed Edge Polyester Cleanroom Wipes
Supplier: High-Tech Conversions
ULTIMATE 60®
ULT60-1212
77008-066CS 516.94 USD InStock
77008-066
ULTIMATE 60™ Sealed Edge Polyester Cleanroom Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
ULTIMATE 60™ cleanroom wipes are made from knit 100% polyester with four laser cut sealed edges.
  • 100% virgin polyester fibers are continuous with no loose ends
  • Laser cut edges
  • Compatible with virtually any solvent or solution for cleaning and wiping
  • Cleanroom laundered and individially inspected
  • Double bagged, vacuum cleanroom packed
  • Autoclavable
  • Meets USP <797> and USP <800> wipe requirements
ULTIMATE 60™ wipes are manufactured in a cleanroom environment and cleanroom packaged. These single-ply cleanroom wipes are absorbent, abrasion-resistant, and manufactured from 100% no-run polyester.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty