Would you like to log in again?
ESD Cleanroom Frocks
Supplier: Transforming Technologies
TX40FRNB05 TX40FRNB07 TX40FRNB04 TX40FRWH01 TX40FRNB09 TX40FRWH03 TX40FRWH06 TX40FRWH02 TX40FRNB08 TX40FRNB01 TX40FRWH04 TX40FRWH05 TX40FRNB06 TX40FRWH09 TX40FRNB02 TX40FRWH07 TX40FRNB03 TX40FRWH08
77008-092EA 40 USD InStock
77008-092 77008-192 77008-142 77008-098 77008-132 77008-088 77008-198 77008-140 77008-196 77008-130 77008-102 77008-114 77008-166 77008-144 77008-178 77008-148 77008-116 77008-204
ESD Cleanroom Frocks
Workwear
ESD-safe integrated cleanroom wear designed for mobile users in controlled environments.
  • TX4000 ESD Grid Fabric; 99% polyester/1% carbon
  • Raglan sleeves with ESD knit cuffs for body contact
  • Military collar
  • Zipper closure with snaps for easy gowning
  • Seams sewn with conductive threads and grounding tape for reliable panel-to-panel electrical continuity
  • Reusable/launderable
  • ISO cleanroom: ISO 6 (Class 1000)
  • Meets standards required for ANSI/ESD S20.20
The Transforming Technologies' TX4000 ESD Cleanroom Garment System protects the controlled environment from the number one source of contamination and static found in cleanrooms - people. Electrostatic charge is an invisible contaminant that has the potential to degrade or destroy both product and equipment, as well as attract unwanted particles. Controlling these charges on mobile operators with the TX4000 Garment System throughout the controlled environment is essential to a productive manufacturing process.
Certifications: In compliance with IEST-RP-CC052.1, ANSI/ESD S20.20.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty