Would you like to log in again?
ProKlean Surface Cleaner
Supplier: Contec
ProKlean
SBC170PK
77008-218CS 251.76 USD InStock
77008-218
ProKlean Surface Cleaner
Cleaning Agents Detergents and Surface Cleansers
Stabilized hypochlorous acid blended with purified water.
  • Filtered to 0.2 µm under ISO Class 5 airflow in a ISO Class 7 cleanroom ensure the product is free from contamination and particulates
  • Contains no quaternary ammonium or surfactant very low residue, saving time on residue removal
  • No hazard classification means good operator acceptability as no strong odor and only basic PPE is required
  • Not classed as corrosive means it can be used safely in all areas of the cleanroom
Contec® ProKlean is a blend of hypochlorous acid in purified water. It is a ready- to-use cleaner safe enough for any surface in a cleanroom. Contec ProKlean is 0.2 µm filtered, and filled under Grade A unidirectional airflow in a Grade B/ISO Class 5 cleanroom. The sterile product is filled into pre-irradiated components. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms. Ready-to-use so can be applied directly to the surface ensuring even coverage. Leave the surface wet for required contact time. Contec ProKlean is low residue but where residues are an issue they are easily removed with either water or alcohol.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty