Would you like to log in again?
Zymo-Seq™ Cell Free DNA WGBS Library Kit
Supplier: Zymo Research
Zymo-Seq™
D5463 D5462
77020-318EA 5172.75 USD InStock
77020-318 77020-284
Zymo-Seq™ Cell Free DNA WGBS Library Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Prepare robust methyl-seq libraries in as little as three steps.
  • Optimized for small fragment input: Ideal for small and damaged DNA fragments such as cell-free DNA (cfDNA)
Zymo-Seq™ cell free DNA WGBS library kit provides an optimized and reliable workflow for whole genome bisulfite sequencing (WGBS) library preparation from cell-free DNA (cfDNA). This all-inclusive kit features a straightforward procedure capable of preparing high-quality methyl-seq libraries from as little as 5 ng of cfDNA. The process is completed in three basic steps: bisulfite conversion, direct adapter ligation, and index PCR amplification. The initial bisulfite treatment is gentle on cfDNA yet effective in converting any unmodified cytosines into uracil.
Next, the innovative splinted adapters capture and directly ligate the illumina-compatible adapters onto any size DNA fragment, allowing for sequencing of nicked, damaged, and short DNA fragments that may have otherwise been discarded with traditional library prep methods. The direct ligation of the adapters also eliminates the need for second strand synthesis, end repair, and dA tailing steps, thus reducing bias by preserving the integrity of any native methylation that is present on the fragment termini. Finally, the adapter-ligated cfDNA is indexed and amplified via PCR with Zymo-Seq™ UDIs, generating libraries that are ready for sequencing on any illumina instrument.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty