Would you like to log in again?
Quick-16S Plus NGS Library Prep Kit (V4)
Supplier: Zymo Research
Quick-16S Plus
D6430-PS2 D6430-PS3 D6430-PS4 D6430-PS1
77020-306EA 1254 USD InStock
77020-306 77020-310 77020-286 77020-322
Quick-16S Plus NGS Library Prep Kit (V4)
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Quick-16S Plus NGS Library Prep Kit (V4) is the fastest and simplest NGS library prep targeting the V4 region of the 16S rRNA gene for high-throughput sequencing.
  • Fast: 8X less hands-on time than conventional 16S library prep protocols
The Quick-16S Plus NGS Library Prep Kit (V4) is the fastest and simplest NGS library prep targeting the V4 region of the 16S rRNA gene for high-throughput sequencing. The automation-friendly protocol utilizes a premixed amplification plate and a single qPCR/PCR for combined targeted amplification and barcode addition. Pooling by equal volume and a single clean-up of the final library, rather than massive AMPure® bead-based clean-ups, simplifies the library prep process. Additional library quantification analysis such as TapeStation® analysis or gel electrophoresis are not necessary. With these features, the workflow dramatically reduces the hands-on time of library preparation to only 30 minutes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty