Would you like to log in again?
Zymo-Spin VP-S Column
Supplier: Zymo Research
Zymo-Spin VP-S
C1083-5
77020-308PK 75.87 USD InStock
77020-308
Zymo-Spin VP-S Column
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The versatile Zymo-Spin V-PS with 15 and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA.
Available as a refill for the ZymoPURE™ II Plasmid Midiprep and ZymoPURE™ – Express Plasmid Midiprep Kits. The versatile Zymo-Spin V-PS with 15 ml and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin V-PS features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 1.2 mg of plasmid DNA in ≥150 µl eluate when used in combination with ZymoPURE™ Plasmid Prep buffers. Capacity with reservoir assembly is 65 ml.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty