Would you like to log in again?
ZymoBIOMICS 96 MagBead DNA Kit
Supplier: Zymo Research
ZymoBIOMICS 96 MagBead
D4302-E D4308-E D4306-E
77020-326EA 1071.34 USD InStock
77020-326 77020-266 77020-296
ZymoBIOMICS 96 MagBead DNA Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Inhibitor-free microbial DNA from any sample validated for microbiome and metagenomic analyses.
  • High-throughput purification of high quality, inhibitor-free DNA from any sample including feces, soil, water, biofilms, swabs, saliva, and body fluids
The ZymoBIOMICS DNA kits are designed for purifying DNA from a variety of sample inputs that is immediately ready for microbiome or metagenome analyses. The ZymoBIOMICS lysis system eliminates bias associated with unequal lysis efficiencies of different organisms (e.g. gram-negative/positive bacteria, fungus, protozoans, and algae), making it ideal for microbial community profiling. Uniform mechanical lysis of all microbes is achieved by bead beating with the innovative ultra-high density bashingbeads. This kit is equipped with our OneStep PCR inhibitor removal technology, enabling PCR amplification from DNA derived from inhibitor-rich environmental samples. Purified DNA is ideal for all downstream applications including PCR, arrays, 16S rRNA gene sequencing, and shotgun sequencing. DNA Size is 15-20 kb.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty