Would you like to log in again?
PTS Panels® Triglycerides Test Strips
Supplier: PTS Diagnostics
PTS Panels®
1716
77020-344EA 135.27 USD InStock
77020-344
PTS Panels® Triglycerides Test Strips
Clinical Diagnostic Supplies Clinical Reagents and Kits
PTS Panels Triglycerides test strips provide lab-quality triglyceride results on-site in about 45 seconds from a single fingerstick.
  • Measures Triglycerides
  • 25 test strips per vial – Professional
  • Refrigeration not required
PTS Panels Triglycerides test strips are for professional use with CardioChek PA and CardioChek Plus analyzers to measure triglycerides in venous whole blood or capillary fingerstick blood.
Triglycerides measurements are used in the diagnosis and treatment of patients with diabetes mellitus, nephrosis, liver obstruction, other diseases involving lipid metabolism, or various endocrine disorders.
This test system will detect triglycerides levels from 50 to 500 mg/dl (0.56 to 5.65 mmol/L) and will display a number value for results in this range.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty