Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
NEBNext® FFPE DNA Library Preparation Kits
Supplier: New England Biolabs
NEBNext®
E6650L E6650S
77025-084EA 815 USD InStock
77025-084 77025-076
NEBNext® FFPE DNA Library Preparation Kits
Nucleic Acid Reagents Next Generation Sequencing Reagents
This kit is designed for use with pre-sheared FFPE DNA.
  • Includes FFPE DNA repair reagents plus optimized library prep reagents and protocol
  • Increased library yields
  • Improved sequencing metrics
  • Greater sensitivity of somatic variant calling
  • Automation-friendly workflow
The NEBNext FFPE DNA Library Prep Kit includes the NEBNext FFPE DNA Repair v2 Mix, an optimized cocktail of enzymes designed to repair FFPE DNA, library prep reagents featuring a new polymerase master mix, and a protocol optimized for FFPE DNA. This kit is designed for use with pre-sheared FFPE DNA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty