Would you like to log in again?
NEBridge Ligase Master Mix, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NEBridge
M1100S M1100L M1100L M1100S
76538-670EA 93 USD InStock
CA76538-670 CA77025-080 76538-670 77025-080
NEBridge Ligase Master Mix, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
NEBridge Ligase Master Mix is a 3X master mix for Golden Gate Assembly. Designed for use with NEB Type IIS restriction enzymes, this master mix contains T4 DNA Ligase in an optimized reaction buffer with a proprietary ligation enhancer.
  • Optimized for efficient and accurate Golden Gate Assembly
  • Convenient 3X Master Mix formatUse with NEB Type IIS Restriction Enzymes
  • Use for seamless cloning –no scar remains following assembly
  • Ideal for ordered assembly of multiple fragments (2 to 50+) in a single reaction
Users need only choose their preferred NEB Type IIS restriction enzyme and add DNA substrates to be assembled. Low complexity single fragment insertions, as well as moderate complexity (3 to 6 fragment) and high complexity (7 to 25 fragment) assemblies, are all supported with this optimized reagent and accompanying protocols.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty