Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Super Erecta® EZ-ADD Wire Shelving

Supplier: Metro International
Super Erecta®
EZA-2460NC EZA-2448NK3 EZA-1860NC EZA-1848NK3 EZA-2436NK3 EZA-1848NC EZA-2436NC EZA-1860NK3 EZA-1836NC EZA-1836NK3 EZA-2448NC EZA-2460NK3
77037-716EA 162.93 USD InStock
77037-716 77037-726 77037-714 77037-724 77037-718 77037-728 77037-730 77037-712 77037-722 77037-710 77037-732 77037-720
Super Erecta® EZ-ADD Wire Shelving
Furniture Shelves
Quickly add more storage to Metro™ wire shelving with Super Erecta® EZ-ADD Wire Shelves. The EZ-ADD wire shelf is designed to be easily added to an existing Metro™ wire shelving unit without dismantling the unit, minimizing the time and frustration associated with reconfiguring shelving.

  • Compatible with Super Erecta, Super Adjustable Super Erecta, and Super Erecta Pro shelving
  • Add/Remove an EZ-ADD shelf easily without dismantling a shelving unit
  • Add more storage levels faster than conventional shelving
  • Built-in Microban antimicrobial product protection fights the growth of bacteria, mold, and mildew (Metroseal Green finish only)
  • Load capacity 181 kg (400 lbs.) evenly distributed
  • Can be used in many different applications, such as stationary and mobile shelving, security shelving, and track shelving solutions
  • Metroseal finish has a 12-year limited warranty against rust and corrosion

The one-piece polymer collar has a built-in bead that captures the groove in a 25 mm (1") diameter Super Erecta® SiteSelect post. A stainless lock plate ensures a tight fit for the collar assembly and holds the shelf in position. Unit load ratings are dependent on the number of Super Erecta® shelves, Super Adjustable shelves, Super Erecta Pro shelves, and/or three-sided double snake frames used. Please see spec sheet for additional details.

Certifications: NSF listed.

Accessories information: Replacement hardware kit includes 4 collars and 4 lockplates. Hardware is included with the purchase of an EZ-ADD shelf.

Ordering information: Metroseal finish has a 12-year limited warranty against rust and corrosion.

Packaging: Single packs consist of 1 EZ-ADD wire shelf, 4 EZ-ADD collars, and 4 lock plates.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty