Would you like to log in again?
CryoFreeze® Cryogenic Vials
Supplier: Scientific Specialties
CryoFreeze®
6131-S0S340AV 6222-S0S350AV 6242-S0S330AV 6122-S0S350AV 6141-S0S330AV 6132-S0S340AV 6121-S0S350AV 6212-S0S350AV 6221-S0S350AV 6142-S0S330AV 6112-S0S350AV
77093-428CS 76.1 USD InStock
77093-428 77093-438 77093-426 77093-436 77093-446 77093-434 77093-444 77093-432 77093-442 77093-430 77093-440
CryoFreeze® Cryogenic Vials
Vials
CryoFreeze® cryo vials offer a secure and convenient environment for your samples. Total quality assurance is central to the design and manufacturing philosophy. The CryoFreeze line brings a new dimension to ultra-low temperature storage.
  • Ideal for vapor-phase liquid nitrogen freezing
  • White writing area with black 100 µl graduations
  • Cap inserts are color coded
  • Round base or self standing Stargrip base
  • Sterile, certified free of DNas, RNase, DNA, PCR inhibitors and endotoxins
  • Internally threaded tube caps have a silicon-rubber O-Ring for a secure, leak-proof seal
  • Externally-threaded tube caps are made from HDPE with a secure lip-seal (no O-ring necessary)
Also available with colored caps. Please visit our website or contact your local Avantor sales office.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty