Would you like to log in again?
OPTI-GUARD® 3 mm Guard Columns
Supplier: Optimize Technologies
OPTI-GUARD®
10-02-01495 10-02-01482 10-02-03050 10-02-01494 10-02-01488 10-02-01485 10-02-01491
77100-026EA 412.88 USD InStock
77100-026 77100-016 77100-018 89209-718 77100-020 89521-904 77100-022
OPTI-GUARD® 3 mm Guard Columns
Chromatography Columns
The OPTI-GUARD® 3 mm maintains the tool-free connectivity of the 1 mm, but incorporates a cartridge based format to allow for a larger bed with more capacity.
  • Hand-tight two-part PEEK/ Stainless Steel holder
  • Connecting the holder requires no extra tubing or fittings
  • Automatically adjusts to any manufacturer's column
  • Perfect repeat ZDV connection
  • Minimal impact - no change in efficiency
  • Holder stays plumbed when changing cartridges
  • Use with OPTI-GUARD® 3 mm cartridges only
The two-part holder is designed for use with any analytical column to provide vital protection from dirty samples and strongly retained contaminants. An auto-adjusting stem conforms to any tube stop depth for a perfect zero-dead-volume connections.
The best way to protect an analytical column from fouling is to install a guard column between the injection valve and the analytical column.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty