Would you like to log in again?
OPTI-SOLV® Mini Filters
Supplier: Optimize Technologies
OPTI-SOLV®
10-04-00097 10-04-02768 10-04-00095
89209-702PK 619.41 USD InStock
89209-702 77100-052 77100-054
OPTI-SOLV® Mini Filters
Filters Precolumn Filters and Frits
The OPTI-SOLV® Mini Filter provides low-impact filtration in a package no larger than a finger-tight fitting.
  • Hand-tight - no tools required 
  • Rated to 6,000psi
  • Designed to filter out particles to extend column life 
  • Perfect Zero-Dead-Volume connections every time 
  • Floating stem automatically adjusts to any depth port 
Use the Mini Filter to extend the life of analytical columns or before mass spectrometers as a last line of defense against debris. Perfect zero-dead-volume connections are obtained every time with the patented automatic tube stop depth adjustment. Unique depth-filtering elements allow for greater capacity with no band spreading or loss of performance. The efficient design requires no holder and threads directly into any 10 to 32 port.
The OPTI-SOLV Mini, Micro and Nano filters are easy to use and perform exceptionally well for volume-critical applications.
Delivery information: Includes five disposable cartridges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty