Would you like to log in again?
OPTI-SOLV® OPTI-SOLV® In-Line Kits
Supplier: Optimize Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
OPTI-SOLV®
10-04-00391 10-04-00383 10-04-00390 10-04-02372 10-04-00386 10-04-02371
89521-910PK 935.12 USD InStock
89521-910 77100-070 77100-072 89521-908 77100-066 77100-068
OPTI-SOLV® OPTI-SOLV® In-Line Kits
Filters In-Line Filters
The user-friendly hand-tight and wrench-tight OPTI-SOLV® In-Line filter system are well suited for a number of applications, such as high temperature, high-pressure environments where quick changes are imperative.
  • Rated to 6,000 psi
  • 0.2 µl total swept volume
  • Versatile for any in-line filtration application
  • Prevents damage to downstream components
  • Incorporates Free-Turn® architecture (*Hand-Tight Version)
  • Plumb once - change filter elements without disconnecting from system
The unique swivel design allows frits to be changed without removing the connecting tubing. Just plumb it once and leave it. The frictional forces generated at the piston/seal interface inevitably lead to particulate shedding. Placement of a filtering element between the pump and injector/auto sampler removes these particulates, preventing problems with downstream components and costly downtime.
The unique swivel design allows frits to be changed without removing the connecting tubing. Just plumb it once and leave it.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty