Would you like to log in again?
OPTI-LYNX™ Direct-Connect Union Kit
Supplier: Optimize Technologies
OPTI-LYNX™
11-02226-AB
77100-310EA 784.29 USD InStock
77100-310
OPTI-LYNX™ Direct-Connect Union Kit
Chromatography Supplies Chromatography Spare Parts
If you are already using our OPTI-LYNX™ Guard Column/Pre-Column Filter Kits and want to use OPTI-LYNX™ at the bottom of the column as well, upgrade with this kit.
  • Quick-connect union solution
  • Quarter-turn design rated to 6,000 psi
  • Low Volume/ Low Dispersion interface for volume critical regions
  • True zero-dead-volume connection
  • Hardware is only plumbed once
  • Hand-tight - no tools required
It includes everything you need to equip one end of any analytical column with straight-through quick-connect operation. Can be used alone or in conjunction with other OPTI-LYNX™ products.
Delivery information: Includes: Universal port adapter, holder cap, sealing element tube, sealing element and fitting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty