Would you like to log in again?
OPTI-LYNX™ Direct-Connect Guard Column Holder Kit
Supplier: Optimize Technologies
OPTI-LYNX™
11-02232-AA
77100-320EA 855.76 USD InStock
77100-320
OPTI-LYNX™ Direct-Connect Guard Column Holder Kit
Chromatography Supplies Chromatography Spare Parts
The OPTI-LYNX™ Guard Column is a high-performance, cartridge-based guard system, suitable for use with any 10 to 32 analytical column due to its auto-adjusting ZDV port adapter.
  • For use with OPTI-LYNX™ Guard column cartridges
  • Quarter-turn quick-connect design rated to 6000 psi
  • Low Volume/ Low Dispersion interface for volume critical regions
  • Repeat zero-dead-volume connection to any column
  • Hardware is only plumbed once
  • Hand-tight - no tools required
OPTI-LYNX™ combines effective column protection with the convenience of quick-connect operation. Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX™ utilizes low dispersion component interfaces to keep extra column volume to a minimum: connection paths have a total internal volume of less than 2 μl. You get quick-connect convenience and low dispersion, all in one package.
Delivery information: Includes: Universal port adapter, holder cap, holder tube and fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty