Would you like to log in again?
OPTI-LYNX™ Universal Port Adapter (UPA)
Supplier: Optimize Technologies
OPTI-LYNX™
11-02290-AA
77100-338EA 425.7 USD InStock
77100-338
OPTI-LYNX™ Universal Port Adapter (UPA)
Chromatography Supplies Chromatography Spare Parts
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
  • For use with OPTI-LYNX™ Hardware
  • Provides repeat ZDV connections regardless of the mating port depths
  • Auto-adjusting connection
  • Adapts any 10-32 column or detector port to OPTI-LYNX™ Hardware
  • Modifies your OPTI-LYNX™ Holder from an 'In-Line' to a 'Direct-Connect' version
The universal port adapter with auto-adjusting 10-32 male connector provides a ZDV connection regardless of the mating port depth. Adapts any 10-32 column or detector port to OPTI-LYNX™ hardware. Set up a simple column-switching rig without the need for expensive high-pressure valves - simply install OPTI-LYNX™ universal port adapters on columns you're getting ready to use, and switch columns quickly and easily (pressure should be allowed to dissipate before column is disconnected).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty