Would you like to log in again?
OPTI-LYNX™ Column Plug
Supplier: Optimize Technologies
OPTI-LYNX™
11-02293-AA
77100-340EA 42.88 USD InStock
77100-340
OPTI-LYNX™ Column Plug
Chromatography Supplies Chromatography Spare Parts
Column plugs can be placed on OPTI-LYNX™ bayonet connectors to seal and protect fluid interfaces, packed beds, frits, and more.
  • For use with OPTI-LYNX™ hardware
  • Fits all OPTI-LYNX™ bayonet connectors
  • Seals and protects fluid interfaces and filters/packed beds
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty