Would you like to log in again?
All-In-One Hardware Kits (use with OPTI-LYNX™), Optimize Technologies
Supplier: Optimize Technologies
11-02323-AA 11-02325-AA 11-02325-AB
77100-348EA 1264.39 USD InStock
77100-348 77100-352 83008-172
All-In-One Hardware Kits (use with OPTI-LYNX™), Optimize Technologies
Chromatography Columns
Opti-Lynx inserts can be a filter, guard column, trap cartridge or analytical column.
Since the interacting surfaces are identical, the only variable component is the holder tube. Install a guard column for one application and then switch to a pre-column filter for the next. Use an in-line filter with different porosities and diameters just by changing the insert. Switch out columns in an instant with the Velocity® system or retrofit current columns with the universal port adapter. You can make and break flow paths for multiple detectors, or anywhere else a break in flow is frequently made. Opti-Lynx allows all of this without ever having to change out the basic components.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty