Would you like to log in again?
OPTI-LYNX™ Holder Cap
Supplier: Optimize Technologies
OPTI-LYNX™
11-02333-AA
77100-362EA 299.72 USD InStock
77100-362
OPTI-LYNX™ Holder Cap
Chromatography Supplies Chromatography Spare Parts
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
  • For use with OPTI-LYNX™ hardware
  • Accepts OPTI-LYNX™ holder tubes of any length
  • Connects to ¹/₁₆" outer Ø. tubing with a 10-32 fitting
  • A necessary part of every OPTI-LYNX™ configuration
Switching between modes of pre-column protection is as easy as opening OPTI-LYNX™ hardware. Simply select the correct holder tube for the OPTI-LYNX™ insert you wish to use (a guard column, pre-column filter, or straight through connection without protection) thread it into the holder cap, pop in your insert, you're ready to go.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty