Would you like to log in again?
OPTI-MAX® Check Valves and Replacement Cartridges, Optimize Technologies
Supplier: Optimize Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
OPTI-MAX®
27-56-01330 22-66-02137 27-57-01330 27-38-01959 22-66-02138 27-38-04713 27-38-00680 27-38-01959 27-38-00682 27-59-01330 27-58-01330 27-38-00672
83008-432EA 2640.89 USD InStock
83008-432 77100-700 77100-702 10123-658 83008-434 77100-704 89522-350 77100-706 77100-692 77100-694 10123-660 77100-696
OPTI-MAX® Check Valves and Replacement Cartridges, Optimize Technologies
Valves Non Return Valves
Preassembled check valve cartridges conveniently insert into a reusable housing and function in either the inlet or outlet position
Conversion kits include the necessary check valves and cartridges to convert a system with OEM style check valves to the OPTI-MAX® cartridge check valve system.
The two-piece Free-Turn™ design allows the outside housing thread to spin independently of the tube fitting port so the housing can be removed from the pump head with the tubing attached. A single cartridge is interchangeable across multiple pump brands. Ceramic ball and seat sets are precision-matched to exacting standards.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty