Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EZBio® PETG Titanium Round Media Bottles, GL45 Neck

Supplier: Foxx Life Sciences
EZBio®
507-1931-FLS 507-1831-FLS 507-1A31-FLS 517-1A32-FLS 517-1932-FLS 507-1631-FLS 517-1832-FLS 517-1632-FLS
77106-992CS 103.24 USD InStock
77106-992 77107-000 77106-994 77106-990 77106-996 77106-986 77106-998 77106-988
EZBio® PETG Titanium Round Media Bottles, GL45 Neck
Bottles Plastic Bottles
Foxx Life Sciences new EZBio®pure Titanium Round (TR) media bottles have enhanced the media bottle design. The improved design features an angled shoulder to ensure smooth and complete media pouring.

  • Media bottles available in 125, 250, 500 and 1000 ml sizes
  • Made entirely of 100% USP Class VI components
  • Compatible with leakproof GL45 VersaCaps®, for flexibility of interchangeable connectors
  • Bottles can withstand low temperatures down to −70 ºC
  • Manufactured in a Class 7 cleanroom

The new titanium round media bottles are perfect for storing and sampling active pharmaceutical ingredients, bulk intermediates, and preparing and storing buffers for culture. These bottles can also be used for long-term storage of pH sensitive liquids. Molded finger grips improve the ability to grasp a bottle, even with gloved hands. The lower height enhances stability, and bottles also feature molded graduation marks that are easy to read. EZBio®pure TR Bottles are made of Polyethylene Terephthalate Glycol (PETG) resin. The crystal clear material uses USP Class VI resins.

Certifications: USP Class VI components.

Packaging: Bottles are packaged in shrinked-wrapped modules.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty