Would you like to log in again?
LabFit™ Base Cabinet Drawers with Door
Supplier: TrippNT
LabFit™
53666 53686 53608 53649 53688 53650 53675 53690 53691 53636 53665 53668 53687 53664
77135-308EA 2514.09 USD InStock
77135-308 77135-358 77135-314 77135-328 77135-306 77135-310 77135-354 77135-350 77135-262 77135-276 77135-352 77135-280 77135-360 77135-282
LabFit™ Base Cabinet Drawers with Door
Furniture Casework
TrippNT® LabFit™ casework is constructed of half-inch polypropylene which is ideal for highly corrosive enviroments.
  • Non-metal construction
  • Resistant to chemicals and corrosion
  • Easy to clean and disinfect
  • Large removeable rear access panel
  • Embedded magnetic door latches
  • Industry leading two year full replacement warranty
  • Made in USA
Polypropylene is a versatile plastic material that offers numberous beneftis for controlled environments. Its durability and resistance to corrosion make it a practical choice. It can endure high temperatures and exposure to various chemicals, making it an ideal option for a range of laboratory and medical applications. Joints are welded insuring a leak-tight, flush union. All hinges, handles, drawer slides, and stops are constructed of polypropylene.
Delivery information: TrippNT® LabFit™ laboratory casework has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor furniture specialist.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty