Would you like to log in again?
LabFit™ Base Cabinet Doors
Supplier: TrippNT
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
53667 53680 53677 53682 53678 53679 53681 53670 53630 53669 53671 53672 53659
77135-318EA 2231.81 USD InStock
77135-318 77135-336 77135-338 77135-316 77135-322 77135-332 77135-266 77135-312 77135-334 77135-340 77135-342 77135-320 77135-298
LabFit™ Base Cabinet Doors
Furniture Casework
TrippNT® LabFit™ casework is constructed of half-inch polypropylene which is ideal for highly corrosive enviroments.
  • Non-metal construction
  • Resistant to chemicals and corrosion
  • Easy to clean and disinfect
  • Large removeable rear access panel
  • Embedded magnetic door latches
  • Industry leading two year full replacement warranty
  • Made in USA
Polypropylene is a versatile plastic material that offers numberous beneftis for controlled environments. Its durability and resistnace to corrosion make it a practical choice. It can endure high temperatures and exposure to various chemicals, making it an ideal option for a range of laboratory and medical applications. Joints are welded insuring a leak-tight, flush union. All hinges, handles, drawer slides, and stops are constructed of polypropylene.
Delivery information: TrippNT® LabFit™ laboratory casework has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor furniture specialist.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty