Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylboric acid-2-aminoethyl ester (2-APB) 98%

Supplier: Ambeed
Warning
Synonyms: 2-Aminoethyl diphenylborinate2-((Diphenylboryl)oxy)ethanamine

A682989-5G A682989-25G A682989-1G A682989-10G A682989-1MG
77145-570EA 5.15 USD InStock
77145-570 77145-572 77145-574 77145-566 77145-568
Diphenylboric acid-2-aminoethyl ester (2-APB) 98%
Diphenylboric acid 2-aminoethyl ester (2-APB)
Caution: for Research Use Only


Formula: C₁₄H₁₆BNO
MW: 225.1 g/mol
MDL Number: MFCD00014823
CAS Number: 524-95-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty